4,4-Difluorocyclohexanecarbonitrile 95%

Supplier: Ambeed
Warning

A217264-100MG A217264-1G A217264-250MG A217264-5G
76873-834EA 18 USD
76873-834 76873-836 76873-838 77883-542
4,4-Difluorocyclohexanecarbonitrile 95%
4,4-Difluorocyclohexanecarbonitrile

Precaución: for Research Use Only

Formula: C₇H₉F₂N
MDL Number: MFCD11847832
CAS Number: 922728-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR