4-Bromo-1H-indazole-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A217122-100MG A217122-1G A217122-250MG A217122-5G
76873-826EA 10 USD
76873-826 76873-828 76873-830 76873-832
4-Bromo-1H-indazole-6-carboxylic acid 95%
4-Bromo-1H-indazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MDL Number: MFCD07781800
CAS Number: 885523-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR