3-Bromo-2-isopropoxy-5-methylpyridine 97%

Supplier: Ambeed
Warning

A217117-1G A217117-5G
76873-822EA 56.23 USD
76873-822 76873-824
3-Bromo-2-isopropoxy-5-methylpyridine 97%
3-Bromo-2-isopropoxy-5-methylpyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
MW: 230.102 g/mol
MDL Number: MFCD17926523
CAS Number: 760207-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR