3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenyl)boronic acid

A216496-1G A216496-250MG A216496-5G
76873-814EA 131.56 USD
76873-814 76873-816 76873-818
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 98%
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BN₂O₃S
MW: 250.0800 g/mol
MDL Number: MFCD07363799
CAS Number: 871333-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR