3-Fluoro-5-isopropoxybenzeneboronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-isopropoxyphenylboronic acid, (3-Fluoro-5-isopropoxyphenyl)boronic acid

A215518-1G A215518-250MG A215518-25G A215518-5G
76873-802EA 57 USD
76873-802 76873-804 77883-529 77883-530
3-Fluoro-5-isopropoxybenzeneboronic acid ≥95%
3-Fluoro-5-isopropoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₃
MDL Number: MFCD07363781
CAS Number: 1195945-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR