tert-Butyl-3-amino-3-methylazetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-amino-3-methylazetidine-1-carboxylate

A215323-100MG A215323-1G A215323-250MG A215323-5G
76873-794EA 34 USD
76873-794 76873-796 76873-798 76873-800
tert-Butyl-3-amino-3-methylazetidine-1-carboxylate 97%
tert-Butyl-3-amino-3-methylazetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD12408556
CAS Number: 1158758-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR