4'-Cyano-2,2,2-trifluoroacetophenone 95%

Supplier: Ambeed
Warning

A214396-100MG A214396-1G A214396-250MG
76873-768EA 44 USD
76873-768 76873-770 76873-772
4'-Cyano-2,2,2-trifluoroacetophenone 95%
4'-Cyano-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₄F₃NO
MDL Number: MFCD01319971
CAS Number: 23516-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR