1-Boc-6-cyanoindole-3-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A213688-1G A213688-100MG A213688-250MG A213688-5G
76873-750EA 67 USD
76873-750 77883-523 77883-524 77883-525
1-Boc-6-cyanoindole-3-boronic acid pinacol ester 97%
1-Boc-6-cyanoindole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₅BN₂O₄
MDL Number: MFCD15143605
CAS Number: 1218790-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR