2-(5-Chloro-4-methylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Ambeed
Warning

A213401-100MG A213401-1G A213401-250MG A213401-5G
76873-742EA 36.08 USD
76873-742 76873-744 76873-746 76873-748
2-(5-Chloro-4-methylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(5-Chloro-4-methylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₁H₁₆BClO₂S
MW: 258.57 g/mol
MDL Number: MFCD12405478
CAS Number: 1111095-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR