Ethyl piperazine-2-carboxylate dihydrochloride 97%

Supplier: Ambeed
Warning

A213303-250MG A213303-5G
76873-740EA 18.04 USD
76873-740 77550-374
Ethyl piperazine-2-carboxylate dihydrochloride 97%
Ethyl piperazine-2-carboxylate dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂O₂
MW: 231.12 g/mol
MDL Number: MFCD22381964
CAS Number: 129798-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR