4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) 95%

Supplier: Ambeed
Warning

A212806-1G A212806-250MG A212806-5G
76873-724EA 27 USD
76873-724 76873-726 76873-728
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) 95%
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO)

Precaución: for Research Use Only

Formula: C₁₀H₂₀NO₂
MDL Number: MFCD00270334
CAS Number: 95407-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR