5-(Chlorosulfonyl)-2-ethoxybenzoic acid 95%
Supplier: Ambeed
|
Danger
|
A212513-10G
A212513-1G
A212513-250MG
A212513-25G
A212513-5G
76873-708EA
380.87
USD
76873-708
76873-710
76873-712
76873-714
76873-716
5-(Chlorosulfonyl)-2-ethoxybenzoic acid 95%
5-(Chlorosulfonyl)-2-ethoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₉ClO₅S MW: 264.68 g/mol |
MDL Number:
MFCD09701452 CAS Number: 200575-16-2 UN: 3261 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...