1-Bromo-4,5-dichloro-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A212451-1G A212451-250MG A212451-25G A212451-5G A212451-10G
76873-700EA 17 USD
76873-700 76873-702 76873-704 76873-706 77883-510
1-Bromo-4,5-dichloro-2-nitrobenzene 97%
1-Bromo-4,5-dichloro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂NO₂
MDL Number: MFCD07780670
CAS Number: 93361-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR