7-Bromo-1H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Danger

A212196-100MG A212196-5G
76873-696EA 40.32 USD
76873-696 77612-084
7-Bromo-1H-benzo[d][1,2,3]triazole 95%
7-Bromo-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
MDL Number: MFCD11226749
CAS Number: 1064721-11-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR