1-Acetyl-4-(5-bromopyridin-2-yl)piperazine 98%

Supplier: Ambeed
Warning

A211142-1G A211142-250MG A211142-5G
76873-688EA 62.99 USD
76873-688 76873-690 76873-692
1-Acetyl-4-(5-bromopyridin-2-yl)piperazine 98%
1-Acetyl-4-(5-bromopyridin-2-yl)piperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN₃O
MDL Number: MFCD11954763
CAS Number: 494771-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR