2-Aminofuran, N-BOC protected 97%

Supplier: Ambeed
Warning

Synonyms: 2-[(tert-Butoxycarbonyl)amino]furan, 2-(Boc-amino)furan, tert-Butyl (fur-2-yl)carbamate

A211083-1G A211083-250MG A211083-5G
76873-682EA 29.71 USD
76873-682 76873-684 76873-686
2-Aminofuran, N-BOC protected 97%
2-Aminofuran, N-BOC protected

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃
MW: 183.204 g/mol
MDL Number: MFCD02682023
CAS Number: 56267-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR