(S)-2-(Methylamino)-3-phenylpropan-1-ol 97%

Supplier: Ambeed
Danger

A210690-100MG A210690-1G A210690-250MG
76873-662EA 45.62 USD
76873-662 76873-664 76873-666
(S)-2-(Methylamino)-3-phenylpropan-1-ol 97%
(S)-2-(Methylamino)-3-phenylpropan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MW: 165.232 g/mol
MDL Number: MFCD18431682
CAS Number: 84773-29-5
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR