2-Bromo-1-(4-ethylphenyl)ethanone 95%

Supplier: Ambeed
Danger

A210411-100MG A210411-1G A210411-250MG A210411-5G
76873-648EA 20.16 USD
76873-648 76873-650 76873-652 76873-654
2-Bromo-1-(4-ethylphenyl)ethanone 95%
2-Bromo-1-(4-ethylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
MW: 227.10 g/mol
MDL Number: MFCD00981997
CAS Number: 2632-14-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR