Methyl 3-amino-2,4-dimethylbenzoate 96%

Supplier: Ambeed
Warning

A209779-1G A209779-5G A209779-250MG
76873-632EA 158.08 USD
76873-632 76873-634 77671-860
Methyl 3-amino-2,4-dimethylbenzoate 96%
Methyl 3-amino-2,4-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD22494937
CAS Number: 24812-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR