Ethyl-(S)-2-amino-4-phenylbutanoate hydrochloride 98%

Supplier: Ambeed
Warning

A208611-100G A208611-10G A208611-25G A208611-5G
76873-624EA 60 USD
76873-624 76873-626 76873-628 77883-501
Ethyl-(S)-2-amino-4-phenylbutanoate hydrochloride 98%
Ethyl-(S)-2-amino-4-phenylbutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂·HCl
MDL Number: MFCD00190691
CAS Number: 90891-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR