2-Aminooxazol-4(5H)-one 95%

Supplier: Ambeed
Warning

A208238-1G A208238-250MG A208238-5G A208238-10G
76873-618EA 61 USD
76873-618 76873-620 76873-622 77883-499
2-Aminooxazol-4(5H)-one 95%
2-Aminooxazol-4(5H)-one

Precaución: for Research Use Only

Formula: C₃H₄N₂O₂
MDL Number: MFCD00807941
CAS Number: 17816-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR