Pyrrole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A208169-100MG A208169-1G A208169-250MG A208169-5G A208169-25G A208169-100G
76873-610EA 5 USD
76873-610 76873-612 76873-614 76873-616 77567-996 77885-506
Pyrrole-3-carboxylic acid 98%
Pyrrole-3-carboxylic acid

Precaución: For research use only.

Formula: C₅H₅NO₂
MDL Number: MFCD00800594
CAS Number: 931-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR