5-Chloro-2-hydroxy-3-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A207554-250MG A207554-5G A207554-1G
76873-598EA 9 USD
76873-598 76873-600 77612-042
5-Chloro-2-hydroxy-3-methoxybenzaldehyde 95%
5-Chloro-2-hydroxy-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MDL Number: MFCD01103568
CAS Number: 7740-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR