(1-(Phenylsulfonyl)-1H-indol-4-yl)boronic acid 95%

Supplier: Ambeed
Warning

A206534-1G A206534-250MG A206534-5G
76873-580EA 98 USD
76873-580 76873-582 76873-584
(1-(Phenylsulfonyl)-1H-indol-4-yl)boronic acid 95%
(1-(Phenylsulfonyl)-1H-indol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BNO₄S
MW: 301.13 g/mol
MDL Number: MFCD12911912
CAS Number: 1256358-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR