2,6-Dichloro-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A206428-100MG A206428-250MG
76873-576EA 55 USD
76873-576 76873-578
2,6-Dichloro-3-methoxybenzoic acid 98%
2,6-Dichloro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
MDL Number: MFCD23701500
CAS Number: 32890-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR