5-(Hydroxymethyl)-2-(trifluoromethyl)-1,3-thiazole 97%

Supplier: Ambeed
Warning

A205959-1G A205959-5G
76873-572EA 260.99 USD
76873-572 76873-574
5-(Hydroxymethyl)-2-(trifluoromethyl)-1,3-thiazole 97%
5-(Hydroxymethyl)-2-(trifluoromethyl)-1,3-thiazole

Precaución: for Research Use Only

Formula: C₅H₄F₃NOS
MDL Number: MFCD18632535
CAS Number: 131748-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR