ABT-263 98%

Supplier: Ambeed
Warning

Synonyms: (R)-4-(4-((4'-Chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((4-((4-morpholino-1-(phenylthio)butan-2-yl)amino)-3-((trifluoromethyl)sulphonyl)phenyl)sulphonyl)benzamide, Navitoclax

A204180-10MG A204180-1MG A204180-25MG A204180-50MG A204180-100MG A204180-1G A204180-250MG A204180-5MG
76873-512EA 130 USD
76873-512 76873-514 76873-516 76873-518 77671-804 77671-806 77883-477 77883-478
ABT-263 98%
ABT-263

Precaución: for Research Use Only

Formula: C₄₅H₅₅ClF₃N₅O₆S₃
MW: 974.61 g/mol
MDL Number: MFCD12756219
CAS Number: 923564-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR