2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine 98%

Supplier: Ambeed
Warning

A203882-100G A203882-10G A203882-1G A203882-250MG A203882-25G A203882-5G
76873-490EA 418 USD
76873-490 76873-492 76873-494 76873-496 76873-498 76873-500
2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine 98%
2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine

Precaución: for Research Use Only

Formula: C₁₇H₁₉ClN₂O
MDL Number: MFCD13184723
CAS Number: 122368-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR