Sulfamide 97%

Supplier: Ambeed
Warning

A203568-1000G A203568-100G A203568-10G A203568-25G A203568-500G
76873-468EA 127 USD
76873-468 76873-470 76873-472 76873-474 76873-476
Sulfamide 97%
Sulphamide

Precaución: for Research Use Only

Formula: H₄N₂O₂S
MDL Number: MFCD00011606
CAS Number: 7803-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR