N-(4-Chlorophenyl)cyclohexanecarboxamide 97%

Supplier: Ambeed
Warning

A203307-1G A203307-250MG
76873-464EA 19 USD
76873-464 76873-466
N-(4-Chlorophenyl)cyclohexanecarboxamide 97%
N-(4-Chlorophenyl)cyclohexanecarboxamide

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNO
MDL Number: MFCD00448186
CAS Number: 142810-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR