2-Hydroxy-2-methylpropiophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Benzoyl-2-propanol, 2-Hydroxy-2-methyl-1-phenylpropan-1-one
A201838-1000G
A201838-100G
A201838-10G
A201838-500G
A201838-25G
76873-436EA
42
USD
76873-436
76873-438
76873-440
76873-442
77086-058
2-Hydroxy-2-methylpropiophenone 98%
2-Hydroxy-2-methylpropiophenone
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ MW: 164.2000 g/mol |
MDL Number:
MFCD00040710 CAS Number: 7473-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...