(4-Imidazol-1-ylphenyl)methanol 98%

Supplier: Ambeed
Warning

A201419-1G A201419-250MG A201419-25G A201419-5G
76873-410EA 12 USD
76873-410 76873-412 76873-414 76873-416
(4-Imidazol-1-ylphenyl)methanol 98%
(4-Imidazol-1-ylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD02682059
CAS Number: 86718-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR