5-Ethoxypyrimidin-2-amine 97%

Supplier: Ambeed
Warning

A201283-250MG A201283-1G
76873-408EA 57 USD
76873-408 77883-461
5-Ethoxypyrimidin-2-amine 97%
5-Ethoxypyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MDL Number: MFCD00223638
CAS Number: 39268-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR