8-Bromo-6-fluoroquinoline 97%

Supplier: Ambeed
Warning

A201090-250MG A201090-5G A201090-1G
76873-394EA 71.09 USD
76873-394 76873-396 77671-790
8-Bromo-6-fluoroquinoline 97%
8-Bromo-6-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.0451 g/mol
MDL Number: MFCD09907837
CAS Number: 22960-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR