(4R,5R,6S)-4-Nitrobenzyl 3-((diphenoxyphosphoryl)oxy)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: beta-Methyl vinyl phosphate (MAP)

A200530-10G A200530-1G A200530-5G A200530-250MG
76873-376EA 84.88 USD
76873-376 76873-378 76873-380 77611-940
(4R,5R,6S)-4-Nitrobenzyl 3-((diphenoxyphosphoryl)oxy)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate 97%
(4R,5R,6S)-4-Nitrobenzyl 3-((diphenoxyphosphoryl)oxy)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate

Precaución: for Research Use Only

Formula: C₂₉H₂₇N₂O₁₀P
MW: 594.50 g/mol
MDL Number: MFCD01318092
CAS Number: 90776-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR