2-Bromo-4-chloro-1-(difluoromethoxy)benzene 97%

Supplier: Ambeed
Warning

A200327-1G A200327-250MG A200327-25G
76873-372EA 22.28 USD
76873-372 76873-374 77593-298
2-Bromo-4-chloro-1-(difluoromethoxy)benzene 97%
2-Bromo-4-chloro-1-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄BrClF₂O
MW: 257.45 g/mol
MDL Number: MFCD11847190
CAS Number: 1214348-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR