tert-Butyl (1-(4-bromophenyl)cyclopropyl)carbamate 97%

Supplier: Ambeed
Warning

A200217-10G A200217-1G A200217-250MG A200217-5G
76873-354EA 123 USD
76873-354 76873-356 76873-358 76873-360
tert-Butyl (1-(4-bromophenyl)cyclopropyl)carbamate 97%
tert-Butyl (1-(4-bromophenyl)cyclopropyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₂
MW: 312.20 g/mol
MDL Number: MFCD16660261
CAS Number: 360773-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR