(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A199900-1G A199900-250MG
76873-350EA 111.4 USD
76873-350 76873-352
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid 95%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₃₀BNO₆
MW: 391.26 g/mol
MDL Number: MFCD03412130
CAS Number: 216439-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR