H-D-Phg-OtBu 97%

Supplier: Ambeed
Warning

A199712-10G A199712-1G A199712-5G
76873-340EA 65.78 USD
76873-340 76873-342 76873-344
H-D-Phg-OtBu 97%
H-D-Phg-OtBu

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.273 g/mol
MDL Number: MFCD04114053
CAS Number: 65715-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR