4-Bromo-3-methyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-methylpyrazole

A199689-100G A199689-10G A199689-1G A199689-25G A199689-5G
76873-330EA 109.28 USD
76873-330 76873-332 76873-334 76873-336 76873-338
4-Bromo-3-methyl-1H-pyrazole 97%
4-Bromo-3-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅BrN₂
MW: 161.01 g/mol
MDL Number: MFCD00005241
CAS Number: 13808-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR