4-[(Dimethylcarbamoyl)amino]benzeneboronic acid, pinacol ester 95+%

Supplier: Ambeed
Warning

Synonyms: 1,1-Dimethyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea, 4-(3-Dimethylureido)benzeneboronic acid, pinacol ester

A199460-1G A199460-250MG A199460-5G
76873-320EA 118.83 USD
76873-320 76873-322 76873-324
4-[(Dimethylcarbamoyl)amino]benzeneboronic acid, pinacol ester 95+%
4-[(Dimethylcarbamoyl)amino]benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BN₂O₃
MW: 290.166 g/mol
MDL Number: MFCD08689520
CAS Number: 874290-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR