N-(2-Hydroxyethyl)-4-methylbenzenesulfonamide 95+%

Supplier: Ambeed
Warning

A198857-1G A198857-250MG
76873-318EA 140.04 USD
76873-318 77671-760
N-(2-Hydroxyethyl)-4-methylbenzenesulfonamide 95+%
N-(2-Hydroxyethyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃S
MW: 215.27 g/mol
MDL Number: MFCD00464177
CAS Number: 14316-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR