(R)-1-Methylpyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A197688-1G A197688-250MG A197688-25G A197688-5G
76873-286EA 45.62 USD
76873-286 76873-288 76873-290 76873-292
(R)-1-Methylpyrrolidine-2-carboxylic acid 97%
(R)-1-Methylpyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.15 g/mol
MDL Number: MFCD15147029
CAS Number: 58123-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR