6-Fluoro-2-hydrazinylbenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A196956-1G A196956-10G A196956-250MG
76873-264EA 30.01 USD
76873-264 77883-424 77883-425
6-Fluoro-2-hydrazinylbenzo[d]thiazole 97%
6-Fluoro-2-hydrazinylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₆FN₃S
MDL Number: MFCD04448803
CAS Number: 78364-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR