4,5-Dibromo-1,2-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Diamino-4,5-dibromobenzene, 4,5-Dibromobenzene-1,2-diamine

A196403-10G A196403-1G A196403-250MG A196403-25G A196403-5G A196403-100G A196403-500G
76873-244EA 22 USD
76873-244 76873-246 76873-248 76873-250 76873-252 77611-894 77671-748
4,5-Dibromo-1,2-phenylenediamine 98%
4,5-Dibromo-1,2-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₆Br₂N₂
MW: 265.94 g/mol
MDL Number: MFCD00795598
CAS Number: 49764-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR