2-Amino-5-ethylphenol hydrochloride 98%

Supplier: Ambeed
Warning

A194851-10G A194851-1G A194851-25G A194851-5G
76873-200EA 101 USD
76873-200 76873-202 76873-204 76873-206
2-Amino-5-ethylphenol hydrochloride 98%
2-Amino-5-ethylphenol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO
MDL Number: MFCD13181598
CAS Number: 149861-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR