tert-Butyl 3-amino-5-methyl-1H-pyrazole-1-carboxylate 97%

Supplier: Ambeed
Warning

A194750-100MG A194750-1G A194750-250MG
76873-194EA 37.14 USD
76873-194 76873-196 76873-198
tert-Butyl 3-amino-5-methyl-1H-pyrazole-1-carboxylate 97%
tert-Butyl 3-amino-5-methyl-1H-pyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₅N₃O₂
MW: 197.23 g/mol
MDL Number: MFCD13252375
CAS Number: 578008-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR