1-Bromo-2,4-dichloro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A194622-1G A194622-5G
76873-180EA 30.77 USD
76873-180 76873-182
1-Bromo-2,4-dichloro-5-nitrobenzene 98%
1-Bromo-2,4-dichloro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂NO₂
MW: 270.9000 g/mol
MDL Number: MFCD16038032
CAS Number: 65001-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR