4-Hydroxy-2-(trifluoromethyl)benzaldehyde 96%

Supplier: Ambeed
Warning

A194493-10G A194493-1G A194493-250MG A194493-25G A194493-5G
76873-170EA 179 USD
76873-170 76873-172 76873-174 76873-176 76873-178
4-Hydroxy-2-(trifluoromethyl)benzaldehyde 96%
4-Hydroxy-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MDL Number: MFCD15528830
CAS Number: 1243395-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR