2,3-Dibromoanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dibromo-3-methoxybenzene, 2,3-Dibromophenyl methyl ether

A194232-250MG A194232-5G A194232-1G
76873-164EA 37.14 USD
76873-164 76873-166 77611-856
2,3-Dibromoanisole 98%
2,3-Dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
MDL Number: MFCD09864854
CAS Number: 95970-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR